J-KEM® Temperature Controller, Model 210, Chemglass
Supplier: Chemglass
CG-3200-01 CG-3200-02 CG-3200-03 CG-3200-100
80062-836EA 1843.11 USD
80062-836 80062-838 80062-840 10034-456
J-KEM® Temperature Controller, Model 210, Chemglass
Controllers Temperature Controllers
Sufficient for heating mantles up to 5L in size along with most laboratory ovens and hot plates
Controller has the ability to display temperature without heating. The Model 210 controls temperature to within ± 0.1 °C of liquids from 1µL to 5L.
Ordering information: Includes 1/8” dia. x 12” long PTFE coated thermocouple, 10 feet coiled extension cord, and a 14/20 PTFE thermocouple adapter.