Needles for Syringes, Chemglass
Supplier: Chemglass
CG-3075-01 CG-3075-02 CG-3075-03 CG-3075-40 CG-3075-41 CG-3075-42
80062-596EA 106.33 USD
80062-596 80062-598 80062-600 80062-640 80062-642 80062-644
Needles for Syringes, Chemglass
Syringe Needles
All needles are made of #304 full hard stainless steel and have a 22º beveled point for septum penetration along with general use.
Delivery information: Removable needles come complete with the PTFE ferrule necessary for assembly.