Polypropylene Solvent Addition Funnel Pack, Chemglass
Supplier: Chemglass
CG-1758-P-01
80061-536EA 156.69 USD
80061-536
Polypropylene Solvent Addition Funnel Pack, Chemglass
Funnels Dropping Funnels
Unbreakable, polypropylene solvent addition funnels are ideal for use with chromatography columns.
The unique closed-end design facilitates gentle addition of solvent to the column. Perforated funnel stem permits solvent to pour down the column wall without disturbing the top layer of silica gel.
Convenient assortment pack consists of five funnels for use with various column sizes.