Filter Adapters, Neoprene, Complete Set, Chemglass
Supplier: Chemglass
CG-1401-01
80061-056EA 80.08 USD
80061-056
Filter Adapters, Neoprene, Complete Set, Chemglass
Supports Support Sleeves
Neoprene filter adapters are designed to fit between the filtering funnel and flask. Eliminates the boring of rubber stoppers and the difficulties of inserting and removing glass stems in stoppers.
  • For use with Buchner funnels, Grooch crucibles, filter tubes, etc.
  • Can be used individually or nesting with adjacent sizes
  • Neoprene withstands autoclaving
Complete set includes sizes 1-7 with I.D. ranging from 17×7 mm to 75×46 mm and O.D. ranging from 21×11 mm to 86×57 mm. Wall thickness ranges from 2.0 to 5.5 mm.