Tefzel® Flangeless Ferrules, Chemglass
Supplier: Chemglass
CG-1164-F-02
80060-478EA 5.04 USD
80060-478
Tefzel® Flangeless Ferrules, Chemglass
Fittings and Connectors
Ferrules are designed to form a zero dead-volume connection.
  • Use with flangeless nut to ensure tubing is held firmly in place
  • Yellow finish
Ferrule is placed on the end of the tubing with the cone pointing toward the nut, flat end towards the PTFE fitting.