Vitamin B12 (cyanocobalamin) ≥95% (dry basis), Calbiochem®, Millipore®

Supplier: MilliporeSigma

6791-5GM
80058-826EA 407.63 USD
80058-826
Vitamin B12 (cyanocobalamin) ≥95% (dry basis), Calbiochem®, Millipore®
Vitamin B12 (cyanocobalamin)

Part of coenzyme needed for the synthesis of proteins, and nucleic acids and for the metabolism of carbohydrates.


  • Plays a role in the synthesis of myelin
  • Used as an antipernicious anemia factor

Formula: C₆₃H₈₈CoN₁₄O₁₄P
Storage Temperature: Ambient
MDL Number: MFCD00151092
CAS Number: 68-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR