Polysorbate 20, Millipore®

Supplier: MilliporeSigma

Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated

655205-250ML
80058-796EA 106.57 USD
80058-796
Polysorbate 20, Millipore®
Polysorbate 20

Non-ionic detergent used for immunoprecipitation and for the solubilization of membrane-bound proteins (CMC: 59 µM; HLB number: 16.7).

Formula: C₅₈H₁₁₄O₂₆
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now

Specification Test Results

HLB Number 16.7

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR