D-(+)-Trehalose dihydrate

Supplier: MilliporeSigma

Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

625625-50GM
80058-758EA 115.76 USD
80058-758
D-(+)-Trehalose dihydrate
D(+)-Trehalose dihydrate
Formula: C₁₂H₂₂O₁₁·2H₂O
MW: 378.33 g/mol
MDL Number: MFCD00071594
CAS Number: 6138-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR