Tauroursodeoxycholate sodium ≥97% (by TLC), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: Sodium 2-((R)-4-((3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonate, Sodium 2-((R)-4-((3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate, Tauroursodeoxycholic acid sodium salt, Sodium tauroursodeoxycholate

580549-1GM 580549-5GM
80058-720EA 268.54 USD
80058-720 80058-722
Tauroursodeoxycholate sodium ≥97% (by TLC), Calbiochem®, Millipore®
Tauroursodeoxycholate sodium

Tauroursodeoxycholic is a detergent useful for the solubilization of lipids and membrane-bound proteins.

Formula: C₂₆H₄₅NNaO₆S
MW: 522.69 g/mol
MDL Number: MFCD00065451
CAS Number: 35807-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR