Suramin hexasodium ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

574625-50MG
80058-690EA 77.03 USD
80058-690
Suramin hexasodium ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Suramin hexasodium

Suramin, Sodium Salt, a reversible and competitive inhibitor of protein tyrosine phosphatases.

Formula: C₅₁H₃₄N₆Na₆O₂₃S₆
Storage Temperature: Refrigerator
MDL Number: MFCD00210217
CAS Number: 129-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR