D-(-)-Sorbitol

Supplier: MilliporeSigma

56755-1KG
80058-652EA 134.17 USD
80058-652
D-(-)-Sorbitol
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MDL Number: MFCD00004708
CAS Number: 50-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR