Saponin, Calbiochem®

Supplier: MilliporeSigma
Warning

558255-100GM
80058-630EA 139.34 USD
80058-630
Saponin, Calbiochem®
Saponin

Saponin is a glycoside used to permeabilize cellular membranes


It exhibits hemolytic activity.

Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR