Polyvinylpyrrolidone

Supplier: MilliporeSigma

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

5295-100GM
80058-576EA 71.54 USD
80058-576
Polyvinylpyrrolidone
Polyvinylpyrrolidone
Formula: (C₆H₉NO)n
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR