L-Phenylalanine ≥98.5% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine

5202-100GM
80058-546EA 167.4 USD
80058-546
L-Phenylalanine ≥98.5% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
L-α-Phenylalanine
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 275…283 °C
Density: 1.34 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR