Nocodazole ≥99% (by HPLC), Calbiochem®

Supplier: MilliporeSigma

487928-10MG
80058-500EA 147.34 USD
80058-500
Nocodazole ≥99% (by HPLC), Calbiochem®
Nocodazole

Nocodazole is a highly specific anti-microtubular agent that promotes tubulin depolymerization and inhibits mitosis.


  • Pale yellow solid

Formula: C₁₄H₁₁N₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD00005588
CAS Number: 31430-18-9

Order Now

Specification Test Results

Purity (HPLC) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR