MOPS-Na, ULTROL®

Supplier: MilliporeSigma

Synonyms: Sodium 4-morpholin-1-ylpropylsulfonate, 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, 3-(N-Morpholino)propanesulfonic acid sodium salt, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt

475899-100GM
80058-432EA 175.16 USD
80058-432
MOPS-Na, ULTROL®
MOPS-Na
Formula: C₇H₁₄NNaO₄S
MW: 231.25 g/mol
MDL Number: MFCD00064350
CAS Number: 71119-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR