γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99% (by titration), ULTROL® biological buffer, Calbiochem®, Millipore®

Supplier: MilliporeSigma

475898-100GM 475898-500GM 475898-1KG
80058-426EA 129.82 USD
80058-426 80058-430 80058-428
γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99% (by titration), ULTROL® biological buffer, Calbiochem®, Millipore®
γ-(N-Morpholino)propanesulphonic acid (MOPS)

Zwitterionic buffer, useful in pH range 6.5-7.9

Formula: C₇H₁₅NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR