Methylene blue, Calbiochem®

Supplier: MilliporeSigma
Warning

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

457250-1GM
80058-364EA 64.97 USD
80058-364
Methylene blue, Calbiochem®
Methylene blue

Inhibitor of soluble guanylate cyclase. Organic spin-trap agent which forms stable adducts with oxygen free radicals in solution; biological staining agent.

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Melting Pt: 180
Density: 1.757 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
EINECS: 200-515-2
Merck Index: 12,06137

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR