L-Methionine ≥98.5% (by assay), Calbiochem®, Millipore®

Supplier: MilliporeSigma

4500-100GM
80058-348EA 135.58 USD
80058-348
L-Methionine ≥98.5% (by assay), Calbiochem®, Millipore®
L-Methionine

L-Methionine, an essential amino acid, useful as a methyl group donor.

Formula: C₅H₁₁NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR