Magnesium chloride hexahydrate

Supplier: MilliporeSigma

Synonyms: Magnesium dichloride

442615-500GM
80058-330EA 104.33 USD
80058-330
Magnesium chloride hexahydrate
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412
Melting Pt: 117
MDL Number: MFCD00149781
CAS Number: 7791-18-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR