L(+)-Lysine monohydrochloride ≥98% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl

4400-1KG 4400-100GM
80058-326EA 170.82 USD
80058-326 80058-324
L(+)-Lysine monohydrochloride ≥98% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
L(+)-Lysine monohydrochloride

L-Lysine hydrochloride, an essential amino acid.

Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS Number: 657-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR