Sodium dodecyl sulfate (SDS), high purity

Supplier: MilliporeSigma

428016-25GM
80058-308EA 116.43 USD
80058-308
Sodium dodecyl sulfate (SDS), high purity
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MDL Number: MFCD00036175
CAS Number: 151-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR