N-Lauroylsarcosine sodium salt

Supplier: MilliporeSigma

428010-5GM
80058-304EA 332.7 USD
80058-304
N-Lauroylsarcosine sodium salt
N-Lauroylsarcosine sodium salt
Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MDL Number: MFCD00042728
CAS Number: 137-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR