Bicine, ULTROL®

Supplier: MilliporeSigma

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

391336-100GM 391336-1KG
80058-224EA 198.67 USD
80058-224 80058-226
Bicine, ULTROL®
Bicine
Formula: C₆H₁₃NO₄
MDL Number: MFCD00004295
CAS Number: 150-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR