Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), ULTROL®

Supplier: MilliporeSigma

391335-100GM 391335-1KG
80058-220EA 200.43 USD
80058-220 80058-222
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), ULTROL®
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)
Formula: C₈H₁₉NO₅
MDL Number: MFCD00002853
CAS Number: 6976-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR