Guanidinium hydrochloride ≥99% (by titrimetric analysis), ULTROL®, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

369075-1KG 369075-500GM 369075-100GM
80058-172EA 644.23 USD
80058-172 80058-174 80058-170
Guanidinium hydrochloride ≥99% (by titrimetric analysis), ULTROL®, Calbiochem®
Guanidinium hydrochloride

Powerful protein denaturant.

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Absorbance (6.0 M, H₂O) 0.2 at 230 nm; 0.03 at 260 nm
Form White solid
Purity ≥99% by titration
Storage +15 to +30 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR