D-(+)-Galactose

Supplier: MilliporeSigma

Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal

3455-100GM
80058-106EA 130.61 USD
80058-106
D-(+)-Galactose
D(+)-Galactose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00151230
CAS Number: 59-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR