ELUGENT™ Non-Ionic Detergent, Calbiochem®
Supplier: MilliporeSigma
324707-100ML
80058-062EA 126.54 USD
80058-062
ELUGENT™ Non-Ionic Detergent, Calbiochem®
Cleaning Agents Decontaminants
Non-ionic detergent composed of a mixture of alkyl glucosides
  • HLB number: 13.1.
  • Form: Clear, viscous brown to yellow liquid
It is useful for solubilization and purification of membrane-bound proteins. Relatively low cost alternative to n-octyl-β-D-glucopyranoside and n-octyl-β-D-maltoside in large-scale solubilizations.