Curcumin (from Curcuma longa) ≥80% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

M-Clarity™ quality level = MQ100

239802-100MG
80057-972EA 63.81 USD
80057-972
Curcumin (from Curcuma longa) ≥80% (by HPLC), Calbiochem®, Sigma-Aldrich®
Curcumin

A cell-permeable and irreversible antitumor and anti-inflammatory agent that acts as an inhibitor of 5-lipoxygenase (IC₅₀ = 8 µM) and cyclooxygenase (IC₅₀ = 52 µM).

Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR