Calcium chloride dihydrate ≥99% ACS, Calbiochem®

Supplier: MilliporeSigma
Warning

208290-1KG
80057-886EA 147.59 USD
80057-886
Calcium chloride dihydrate ≥99% ACS, Calbiochem®
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Form White solid
Hygroscopic Hygroscopic
Solubility H2O (50 mg/ml)
Purity ≥99% by assay
pH 4.5 - 8.5
Storage +15 - +30 °C
Toxicity Irritant
Stability Following reconstitution, store at room temperature (20 °C). Stock solutions are stable for up to 6 months at room temperature


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR