6-Benzylaminopurine

Supplier: MilliporeSigma

200241-1GM
80057-860EA 57.55 USD
80057-860
6-Benzylaminopurine
6-Benzylaminopurine
Formula: C₁₂H₁₁N₅
MDL Number: MFCD00005572
CAS Number: 1214-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR