APO-12

Supplier: MilliporeSigma

178377-1GM
80057-806EA 173.27 USD
80057-806
APO-12
Protein Purification Protein Purification Reagents

Non-ionic detergent useful in two-phase separation and purification of proteins.


  • Aggregation number 2232, CMC 568 mM.
  • M.W. 246.4.
  • CAS No.: 871-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR