APO-10
Supplier: MilliporeSigma
178375-1GM
80057-804EA 173.27 USD
80057-804
APO-10
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Novel non-ionic detergent useful for plasmid DNA isolation. Plasmid DNA isolated using APO-10 does not require the use of phenol/chloroform or proprietary resins. Provides excellent yields of high quality DNA.
  • CAS No.: 2190-95-6