L-Alanine ≥98% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

1250-100GM
80057-774EA 115.68 USD
80057-774
L-Alanine ≥98% (by titrimetric analysis on dry substance), Calbiochem®, Millipore®
L-Alanine

A non-essential amino acid

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189
Melting Pt: 79
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
Merck Index: 13,00203

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR