Thiamine hydrochloride ≥98%, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

5871-100GM
80057-736EA 106.27 USD
80057-736
Thiamine hydrochloride ≥98%, Calbiochem®
Thiamine hydrochloride

Cofactor required for oxidation of carbohydrates and for synthesis of ribose.

Formula: C₁₂H₁₈Cl₂N₄OS
MDL Number: MFCD00012780
CAS Number: 67-03-8

Order Now

Specification Test Results

Form White solid
Purity ≥98% by assay
Storage +15 °C to +30 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR