Tween® 20 (Polysorbate) 10% in aqueous solution, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: Polyoxyethylene sorbitan monolaurate

Packaged under inert gas.

655206-50ML
80057-684EA 198.39 USD
80057-684
Tween® 20 (Polysorbate) 10% in aqueous solution, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®
Tween® 20 (Polysorbate)

Non-ionic detergent used for immunoprecipitation and for the solubilization of membrane-bound proteins.


  • Conductivity: <50 µS/cm

Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now

Specification Test Results

Aldehyde concentration <200 µM
peroxide concentration <200 µM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR