TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®

Supplier: MilliporeSigma
Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

648463-50ML
80057-666EA 270.71 USD
80057-666
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Non-ionic detergent is used for the isolation of membrane proteins.


Contaminants: Aldehyde concentration: <200 µM; peroxide concentration: <200 µM; conductivity: <50 µmhos


Packaging: Packaged under inert gas

Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR