3,3',5-Triiodo-L-thyronine free acid ≥97% (by HPLC), high purity, chromatographically purified, Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

642511-10MG
80057-656EA 172.98 USD
80057-656
3,3',5-Triiodo-L-thyronine free acid ≥97% (by HPLC), high purity, chromatographically purified, Calbiochem®, Sigma-Aldrich®
3,3',5-Triiodo-L-thyronine
Formula: C₁₅H₁₂I₃NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00002593
CAS Number: 6893-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR