Protease (from Streptomyces griseus)
Supplier: MilliporeSigma
53702-10KU 53702-500KU 53702-50KU 53702-250KU 53702-25KU
80601-398EA 108.58 USD
80601-398 80601-404 80601-406 80057-356 80601-402
Protease (from Streptomyces griseus)
Enzymes
A non-specific protease liquefies mucins and digests proteins to free amino acids.
  • Can be used to isolate living chondrocytes
  • Most active at neutral pH, but stable over wide range of pH and temperature
  • Lyophilized form