Oleic acid

Supplier: MilliporeSigma

4954-1GM
80057-190EA 85.59 USD
80057-190
Oleic acid
Oleic acid
Formula: C₁₈H₃₄O₂
MDL Number: MFCD00064242
CAS Number: 112-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR