p-Nitrophenyl phosphate DiTris salt ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Phosphoric acid 4-nitrophenyl ester Bis[tris(hydroxymethyl)aminomethane] salt, 4-Nitrophenyl phosphate Bis(tris) salt, p-Nitrophenyl phosphate DiTris Salt, 4-Nitrophenyl phosphatedi(tris) salt, pNPP 2Tris

487655-5GM
80057-156EA 56.76 USD
80057-156
p-Nitrophenyl phosphate DiTris salt ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
p-Nitrophenyl phosphate DiTris salt

p-Nitrophenyl Phosphate, DiTris Salt, excellent substrate for alkaline phosphatase-based ELISA assays.

Formula: C₆H₆NO₆P
MW: 461.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037009
CAS Number: 68189-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR