Hydroxyurea, Sigma-Aldrich®

Supplier: MilliporeSigma

400046-5GM
80056-742EA 192.28 USD
80056-742
Hydroxyurea, Sigma-Aldrich®
Hydroxyurea

Antineoplastic agent


Blocks DNA replication by inhibiting ribonucleotide reductase. Induces apoptosis in chronic myelogenous leukemia lymphocytes.

Formula: CH₄N₂O₂
MDL Number: MFCD00007943
CAS Number: 127-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR