GENAPOL®, Protein Grade® Detergent, Calbiochem
Supplier: MilliporeSigma
345798-50ML
80056-508EA 280.17 USD
80056-508
GENAPOL®, Protein Grade® Detergent, Calbiochem
Protein Purification Protein Purification Reagents
GENAPOL® Detergents are polyglycol ethers, purified for the isolation of membrane protein.
  • Filter sterilized
  • Storage at 2 to 8 °C
Additionally purified to reduce levels of contaminating peroxides, aldehydes, metals, and salts.