Dexamethasone, Calbiochem®

Supplier: MilliporeSigma

265005-100MG
80056-298EA 196.37 USD
80056-298
Dexamethasone, Calbiochem®
Dexamethasone

Most active and highly stable glucocorticoid


Inhibits the expression of inducible but not constitutive nitric oxide synthase in vascular endothelial cells (IC50=5nM). Enhances active cation transport in aortic smooth muscle cells by stimulating the Na+-K+ pump. Induces apoptosis in human thymocytes.

Formula: C₂₂H₂₉FO₅
MDL Number: MFCD00064136
CAS Number: 50-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR