DAF-2 diacetate 5 mM in DMSO ≥98% (by HPLC) fluorescent indicator, Calbiochem®

Supplier: MilliporeSigma

Synonyms: 2-​(3,​6-​diacetyloxy-​4,​5-​diamino-​9H-​xanthen-​9-​yl)-​benzoic acid, DAF-2 DA, 4,5-Diaminofluorescein diacetate

251505-1MG
80056-264EA 691.82 USD
80056-264
DAF-2 diacetate 5 mM in DMSO ≥98% (by HPLC) fluorescent indicator, Calbiochem®
DAF-2 diacetate

A cell-permeable derivative of DAF-2 (80056-262) that is hydrolyzed to DAF-2 by intracellular esterases


  • A 5 mM (1 mg/450 µl) solution in DMSO
  • Packaged under inert gas


Can be used in fluorescence microscopy to measure real-time changes in nitric oxide levels in vivo. Not available for sale in Japan.

Formula: C₂₄H₁₈N₂O₇
MW: 446.41 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05865101
CAS Number: 205391-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR