Cytarabine ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Cytosine beta-D-Arabinofuranoside, 1-β-D-Arabinofuranosylcytosine, Arabinocytidine, Ara-C, 4-Amino-1-((2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Cytosine arabinoside, 4-Amino-1-beta-D-arabinofuranosyl-2(1H)-pyrimidinone

251010-1GM
80056-258EA 131.01 USD
80056-258
Cytarabine ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Cytarabine

Anticancer, antiviral agent that is especially effective against leukemias.


  • White solid
  • Storage: +2 °C to +8 °C


Anticancer, antiviral agent that is especially effective against leukemias. Induces apoptosis in human myeloid leukemia cells and in rat sympathetic neurons. Induces G1/S phase cell cycle arrest.

Formula: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00066487
CAS Number: 147-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR