Dithiothreitol (DTT, Cleland's reagent) ≥97.00%, Calbiochem®

Supplier: MilliporeSigma

233155-100GM 233155-25GM 233155-10GM
80056-198EA 1722.27 USD
80056-198 80056-204 80056-206 80056-200 80056-202
Dithiothreitol (DTT, Cleland's reagent) ≥97.00%, Calbiochem®
Dithiothreitol (DTT, Cleland's reagent)

It is a protective agent for thiol groups.


  • Packaged under inert gas

Formula: C₄H₁₀O₂S₂
Boiling Pt: ∼125 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash Pt: >110 °C
MDL Number: MFCD00004877
CAS Number: 3483-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility Ethanol or H₂O (30 mg/ml)
Color White
Form Powder
Purity (SH) lodometric titration ≥97.00%
Oxidized dithiothreitol ≤0.50%
Heavy Metals ≤1.0 ppm
Storage and Handling +2°C to +8°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR