Caffeic acid, Calbiochem®

Supplier: MilliporeSigma

Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid

205546-500MG
80055-978EA 112.17 USD
80055-978
Caffeic acid, Calbiochem®
Caffeic acid

A natural dietary compound reported to have anticarcinogenic and anti-inflammatory properties. A selective, non-competitive inhibitor of 5-lipoxygenase (ID50=3.7µM). Inhibits Ca2+-induced LDL oxidation in the initiation phase but acts as a prooxidant in the propagation phase. An irreversible inhibitor of glutathione S-transferases and a non-competitive inhibitor of xanthine oxidase.

Formula: C₉H₈O₄
MW: 180.16 g/mol
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 234…237 °C
MDL Number: MFCD00004392
CAS Number: 331-39-5
EINECS: 206-361-2
Merck Index: 12,01673

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR