N-tert-Butyl-[alpha]-phenylnitrone, Calbiochem®
Supplier: MilliporeSigma
203995-500MG
80055-972EA 112.92 USD
80055-972
N-tert-Butyl-[alpha]-phenylnitrone, Calbiochem®
Cell Culture Supplements and Additives
Most commonly-used free radical trap
An antioxidant that has been shown to act as a protective agent in several experimental models of neurodegenerative disease. Inhibits lipid peroxidation in rat liver microsomes. Also prevents the induction of inducible nitric oxide synthase (iNOS) by reactive oxygen species.