Cacodylic acid sodium salt trihydrate

Supplier: MilliporeSigma

Synonyms: Dimethylarsinic acid sodium salt trihydrate, Sodium cacodylate trihydrate, Dimethylarsonic acid sodium salt trihydrate, Sodium dimethylarsinate trihydrate

205541-100GM
80055-530EA 379.41 USD
80055-530
Cacodylic acid sodium salt trihydrate
Cacodylic acid sodium salt trihydrate
Formula: (CH₃)₂As(O)ONa·3H₂O
MW: 213.94 g/mol
MDL Number: MFCD00149079
CAS Number: 6131-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR